Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hamilton™ Luer Lock Metal Hub Needles, 10 gauge, Metal Hub NDL, 2 in., point style 5
Luer Lock is an industry standard tapered termination utilized by most syringe manufacturers including medical Hypodermic syringes. Luer Lock needles are common because their design is controlled by a series of universal standards which guarantees compatibility between manufacturers.
Supplier: Hamilton™ 773005
This item is not returnable.
View return policy
Description
- Metal hub (nickel plated brass) needles
- Standard needle length is 2 in. / 51 mm
- Available in convenient six packs
- Custom needle lengths and point styles are available
Warranty and Services
1 year
Specifications
Autoclavable | |
10 G | |
2 in. | |
Metal Hub Needles |
Metal (Hub) | |
Luer Lock Syringes | |
51 mm | |
5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction