Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Protection Apron

Resistant to many water-based liquids, greases and oils, and liquid chemical sprays.
Supplier: Kimberly-Clark Professional™ 97790/EMD
This item is not returnable.
View return policy
Description
- Yellow polyethylene-coated fabric
- Ties in back at waist
- 44 in. long (112cm)
- Case of 100
Specifications
Polyethylene-coated Fabric | |
Bound | |
NFPA 99 | |
111.76 cm |
Yellow | |
1.5 mil | |
44 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title