Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ Scott™ Pro Foam Hair and Body Wash
Two products in one – a gentle body wash and a foaming shampoo. Luxury foam is protein-enriched with emollients and other skin conditioning agents. Foams nicely with pleasant fresh scent. Rinses clean without leaving a soapy residue. Great choice for health clubs, gyms, hospitality – anywhere where guests need to use your shower facilities.
Supplier: Kimberly-Clark Professional™ 91553
Description
- 1.0 L cassettes of Scott Pro Foam Body & Hair Wash; 6 cassettes/case
- Versatile (2-in-1) shampoo and body wash is a great choice where guests need to use the shower
- Rinses clean with no soapy residue
- Contains emollients and skin conditioning ingredients
- Use Scott Pro Foam Hair & Body Wash with a compatible Scott Pro dispenser for convenient, consistent dispensing; It’s easy to refill when you buy in bulk
- Fresh, light scent
Specifications
Hair and Body Wash | |
Foam | |
5°C to 37°C | |
2.4 lb. |
Light Blue | |
5 to 6 | |
33.8 oz. | |
0.9 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction