Learn More
Kimberly-Clark Professional™ KleenGuard™ A10 Bouffant Caps
GREENER_CHOICE
Catalog No. 19057600
Click to view available options
Gun metal frame; PC, Indoor/Outdoor lens
This item is not returnable. View return policy
This item is not returnable. View return policy
KleenGuard™ A10 Bouffant Caps provide economical, disposable protection for workers exposed to dirt and grime in the workplace.
  • Protective wear designed for entry-level protection from hazardous dust and feature an elastic opening
  • Breathable fabric is made of lightweight spunbound material providing a cool, comfortable work environment
  • Ideal choice for food processing, general maintenance and manufacturing
  • Not made with natural rubber
  • Elasticized opening
  • A10 fabric is five layers of stacked barrier protection (SMMMS) against hazardous particles, offering economical protection
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bouffant
Size 24 in.
Material Spunbond Polypropylene
Disposable Single-use
Model 36920
For Use With (Application) Food Processing, General Maintenance, and Manufacturing
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.