Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Celestron™ Kids Microscope Kits

Celestron™ Kids Microscope Kits include all of the essential items needed to begin exploring the wonders of the microscopic world
$36.50 - $51.75
Specifications
Product Type | Microscope |
---|---|
Objective Types | 3 objective lenses: 100x, 600x, and 1200x |
For Use With (Application) | Ages 8+ |
Warranty | 2 Years |
Alert:
Warning: Contains Chemicals Listed as Cancer and Reproductive Harm in the State of California
Description
Basic Kit
- Up to 1200x power
- Microscope kit includes 100x, 600x, and 1200x objectives
- Bonus 2x magnifying glass included
- Additional accessories includes 7 blank slides with covers
- CPSIA and ASTM F963 tested and compliant
- Age graded 8+
- Beginner set with metal bodied biological microscope
- 3 objective lenses for high power viewing - 100x, 600x, and 1200x
- Form fitted carry case for portability and protection
- 7 blank slides, 5 prepared specimen slides, and 7 slide cover slips and labels
- Accessories to get you experimenting right away: plastic scalpel, tweezers, probe, 2x magnifier, and specimen samples
- CPSIA and ASTM F963 tested and compliant
- Age graded 8+
Specifications
Microscope | |
3 objective lenses: 100x, 600x, and 1200x | |
Ages 8+ | |
2 Years |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title