Learn More
TARGETMOL CHEMICALS INC HYDROXY-DYNASORE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6822
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hydroxy-Dynasore (Dyngo-4a) a effective dynamin inhibitor inhibits withDynI (brain) DynI (rec) and DynII (rec) of IC50 of 0.38 uM 1.1 uM and 2.3 uM respectively. purity: 98%
Catalog No. 50-257-6442
Explore available promotions