Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ HM 430, HM 450 and 440 E Sliding Microtome Accessories

Maintain Epredia™ HM 430 and HM 450 sliding microtomes with Epredia™ HM 430, HM 450 and 440 E sliding microtomes accessories.

Supplier:  Epredia™ 448070

 View more versions of this product

Catalog No. 14-070-85


Explore available promotions
Add to Cart

Description

Description

Standard buffer tray

  • Holds stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
  • Cooling element for buffer solution

  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
  • Large-field magnifier

  • Magnification 2.5X
  • Move, swivel, and displace for optimal viewing position
  • Includes adapter and illumination
  • StemiScope™ with optics and mount

  • Eyepieces: 10X
  • Supplementary lens: 0.63X
  • Zoom magnifying range: 4.1 to 31.5X
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
  • Swan-neck light guide

  • 3.5 mm fiber bundle
  • Swan-neck light guide: 50 mm long
  • Flexible light guide: 750 mm long
  • High light intensity
  • Include adapter for the KL200 (760317)
  • Specifications

    Specifications

    Height Adjustment Plate, 5 mm
    Sliding Microtome Accessories
    Epredia HM 430 and HM 450 Sliding Microtomes
    Product Suggestions

    Product Suggestions

    Safety and Handling

    Safety and Handling

    WARNING: Cancer - www.P65Warnings.ca.gov
    SDS
    Documents

    Documents

    Product Certifications
    Promotions

    Promotions

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.