Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Guardian™ 3000 Hotplate and Stirrer
SCI_ED

Guardian™ 3000 Hotplate and Stirrer is designed for economical heating and stirring in all laboratory settings. Select from three plate sizes to suit your application. All models can be used with the optional temperature probe for precise temperature control of the sample.

$439.00 - $493.00

Specifications

Speed 80 to 1600 rpm
Voltage 115 V
Controller Type Digital
Frequency 50/60 Hz
Max. Sample Load 15 L
View More Specs

Products 3
Catalog Number Mfr. No. Size Top Plate Material Temperature (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Top Plate Material Temperature (Metric) Price Quantity & Availability  
S81903
SCI_ED
View Documents
OHAUS™
30680266/EMD
135 mm dia. Ceramic-coated Stainless Steel Ambient +5° to 380°C
Each for $439.00
Add to Cart
 
S81902
SCI_ED
View Documents
OHAUS™
30680268/EMD
4 x 4 in. Ceramic Ambient +5° to 500°C
Each for $439.00
Add to Cart
 
S81901
SCI_ED
View Documents
OHAUS™
30680267/EMD
7 x 7 in. Ceramic Ambient +5° to 500°C
Each for $493.00
Add to Cart
 
Description

Description

  • Applications: Boiling Liquids, Water/Oil Bath Preparation, Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
  • Display: Backlit LCD Display for temperature and speed, green indicator lights when unit is heating and stirring, prominent hot top indicator light, probe in use icon, Single Point Calibration icon
  • Operation: Stirring from 80 to 1600 rpm, ambient temperature range from +5°C to 500°C (ceramic top) or 380°C (ceramic-coated steel top), power cord (included), stirring models include PTFE-coated stir bar
  • Communication: N/A
  • Construction: Durable metal housing, PET front panel, control knobs, on/off switch for heater disconnect, built-in support rod holder, easy-to-clean ceramic or ceramic-coated stainless steel tops
  • Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring, temperature probe connector for use with optional temperature probe
Specifications

Specifications

80 to 1600 rpm
Digital
15 L
115 V
50/60 Hz
Custom Backlit LCD
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.