Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Graduated Squibb Separatory Funnels with PTFE key Stopcock

High quality borosilicate 3.3 glass funnel with PTFE key stopcocks and interchangeable polypropylene stoppers
Supplier: Eisco™ CH0479EGR
Description
Warranty and Services
1 year limited manufacturer warranty
Specifications
1000 mL | |
53.3 cm | |
Clear | |
Separating liquids | |
Graduated Squibb-style separatory funnel | |
PTFE Key | |
29/32 |
Borosilicate Glass | |
Yes | |
Yes | |
Funnel, stopper | |
1 | |
Polypropylene |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title