Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,2-Dimethoxyethane | 110-71-4 | MFCD00008502 | 1 L
Supplier: Sigma Aldrich Fine Chemicals Biosciences 2595271L
1,2-Dimethoxyethane | Purity: 99.5% | Mol Wt: 90.12 | 110-71-4 | MFCD00008502 | 1L ** For R&D use only. Not for drug, household, or other uses.