Learn More
Electron Microscopy Sciences DermaShield Skin Protection Lotion 150 ML
SDP
Supplier:  Electron Microscopy Sciences 7057310
Encompass_Preferred
DermaShield Lotion provides a non-toxic and non-greasy invisible layer on all skin areas to protect against most skin abusers, such as harsh soaps and detergents, alcohol, oil, grease, solvents, chemicals, inks, and polishing products. It also provides emergency protection against bases and most acids.
It may reduce incidences of skin allergies and help maintain the skin's natural moisture balance. It gives up to 4 hours maximum protection and withstands repeated washings. DermaShield is safe for all skin areas and allows the skin to breathe and perspire normally. DermaShield Lotion is intended to to be used in addition with conventional protection.
Catalog No. 50-193-0713
Explore available promotions