Learn More
TARGETMOL CHEMICALS INC DEGUELIN 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6817
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Deguelin ((-)-Deguelin) is a PI3K/AKT Inhibitor which is a natural product isolated from plants in the Mundulea sericea family. purity: 98%
Catalog No. 50-257-6444
Explore available promotions