Learn More
Fine Science Tools Castroviejo Micro Needle Holders, Curved, 9 cm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fine Science Tools 1206102
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Product: Castroviejo Micro Needle Holder - Curved/Smooth/9cm/without Lock/ Tip Shape: Curved/ Tip Width: 0.6mm/ Clamping Length: 5mm/ Scissors: No/ Lock: No/ Alloy: Stainless Steel/ Length: 9cm/ Serrated: No/ Unit: Each