Learn More
Zahm & Nagel CO2 Purity Tester, 99-100% in 0.01%
Supplier:  Zahm & Nagel 10003
Encompass_Preferred
The Purity Tester consists of a caustic reservoir and calibrated absorption burette mounted to a PVC polished frame. The reservoir and burette are held to the frame with two neoprene covered mounting bands that are adjustable from the rear side of the frame. The absorption burettes are available in five different calibrations, with the standard burette graduated in tenths of one percent, the total being one percent of the volume of the burette. The caustic reservoir (10100) has a line indicating the level to which caustic solution should be added, which volume is slightly in excess of the capacity of the absorption burette.
Catalog No. NC1320930
Explore available promotions