Learn More
Bevco Precision Manufacturing Co Doral Mid Height ISO 5 Cleanroom Blue Vinyl Chair; Non-Tilt; Polished Aluminum Base w/Adjustable Footring
Supplier:  Bevco Precision Manufacturing Co 5550C2BLU
Encompass_Preferred
BEVCO’s Doral ISO 5 Cleanroom Series models are designed to meet the particulate goals of ISO 14644-1 . A unique filtration system traps particles for contamination-free comfort has been tested by an independent laboratory to certify ISO 5 compliance. Specifically, users that have a petite-to-average build will truly enjoy this seating experience. This non-tilt model features manual back and seat height adjustments for increased comfort during extended sitting durations. Fully adjustable production-style back has a durable scuff resistant back pan to protect the upholstery. A waterfall seat promotes lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 21. 5" - 31. 5". Blue vinyl meets CAL 117 standards. The chair features a sturdy 27" diameter chrome tubular steel base with adjustable chrome footring.
Catalog No. NC2022915
Explore available promotions