Learn More
METTLER TOLEDO CERAMIC DIAPHRAGM CLNR 250ML
Supplier:  METTLER TOLEDO 51340070
Encompass
Thiourea solution for cleaning silver sulfide contamination from diaphragms. Acts to discoloration. Size 250 mL.
Catalog No. NC0260549
Explore available promotions