Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Catalase
SCI_ED

Oxidizing Enzyme

$9.50 - $35.75

Specifications

Concentration 20,000 U/g
Color White to Tan
Purity 100%
For Use With (Application) Laboratory chemicals
Content And Storage Refrigerated
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
S25239A
SCI_ED
View Documents
1 g
Each for $35.75
Edge
Add to Cart
 
S25239
SCI_ED
View Documents
40 mg
Each for $9.50
Add to Cart
 
Description

Description

  • Powder
  • CAS Reg. 9001-05-2
  • Shelf life: 12 months
  • Recommended Storage: Refrigeration

Specifications

Specifications

20,000 U/g
100%
Refrigerated
Powder
White to Tan
Laboratory chemicals
Catalase
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.