Learn More
Motic™ C-mount Camera Adapters
SCI_ED
The C-mount camera adapter allows Moticams to connect with Motic™ trinocular microscopes.
Supplier:  Motic™ 1101001901781
 View more versions of this product
Catalog No. 01-674-239
Explore available promotions
Add to Cart
Description
Description
  • Connects your Moticam with your BA310MET Series Microscope to capture imaging
Specifications
Specifications
Camera Adapter
BA310MET Series 2/3 in. Chip Sensor
0.65X
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov