Learn More
Perkin Elmer US LLC Burner Head Cleaning Tool for AA
Supplier: Perkin Elmer US LLC 03031573
This burner head cleaning tool was designed to clean off any build up from the slots of the Atomic Absorption (AA) burner heads.