Learn More
Omega Scientific Inc Bovine Calf Serum, Heat Inactivated, 500 mL
Supplier: Omega Scientific Inc BC06
Bovine Calf Serum is produced with raw material collected from calves ranging from 14 days and six months of age. 0.2 micron Filtered, Mycoplasma Tested. Primarily US origin, other sources may be available upon request. Virus Screened. Triple 0.1 Micron Filtered. Heat Inactivated. 500 mL
This item is not returnable.
View return policy