Learn More
PIP™ Bouton™ Optical Stone™ Visor for Safety Goggles
Click to view available options
Visor; 1.6mm thick
Description
ANSI rated polycarbonate visor can be attached and removed to Stone Goggle in seconds
- Transparent Gray
- Provides lower face protection
- One Size
Compliance
ANSI rated
Specifications
Description | Visor |
For Use With (Equipment) | Stone safety goggles |
For Use With (Application) | Lower face protection |
Disposable | Reusable |
Lens Material | Polycarbonate |