Learn More
Consolidated Plastics 8 Oz. Bottle - 2 1/4"Dia x 7"H - 24mm CapBoston Round Bottles With Yorker Dispensing Caps
SDP
Supplier:  Consolidated Plastics 41119
Encompass_Preferred
Low Density Polyethylene (LDPE)Boston-Round plastic squeeze bottles are excellent for storing and dispensing a wide variety of products. Color is natural. Yorker dispensing cap included. FDA accepted. Orifice size is .030 in. Height includes cap.
Catalog No. NC1137923
Explore available promotions