Learn More
Med Vet International Boston Round Yorker Dropper Bottle, 60mL/2 oz., 25/bag
Supplier:  Med Vet International BSRD60
Encompass_Preferred
Removable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screwcap made of polystyrene. 25 per bag.
Catalog No. 50-264-3581
Due to product restrictions, please to purchase or view availability for this product.
Explore available promotions