Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Biohazard Disposable Bags
Biohazard bags provide a safe method of disposal for biohazard waste.
Supplier: United Scientific™ UNU40106
Alert:
Prop 65
Description
- Bags have a clearly printed biohazard symbol and pass ASTM 1922 tear resistance and ASTM 1709 dart impact tests.
- Steam autoclavable for sterilization of waste before disposal.
- Polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
- High temperature steam autoclavable to 135°C (275°F) Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests 55 microns (2.2 mil) thickness
Specifications
Autoclavable | |
Red | |
Biohazard Material | |
Non-sterile | |
12 in. |
9 L | |
Single-use | |
Polypropylene | |
24 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Warning: Cancer and reproductive harm
Spot an opportunity for improvement?
Provide Content Correction