Learn More
Bellco Glass, Inc. Bell-Flo iob assembly was designed to be used with the bell-flo vessel with bottom indents and 3 vertical baffles for optimal mixing. Rugged stainless steel ptfe internal bearing impeller assemblies utilize a wing style impeller blade that increases mixin
Supplier: Bellco Glass, Inc. 196580101
Bell-Flo iob assembly was designed to be used with the bell-flo vessel with bottom indents and 3 vertical baffles for optimal mixing. Rugged stainless steel ptfe internal bearing impeller assemblies utilize a wing style impeller blade that increases mixin