Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD PBX Calcium Assay Kit
For homogeneous measurement of intracellular calcium changes
$682.00 - $3980.00
Specifications
For Use With (Application) | For homogeneous measurement of intracellular calcium changes |
---|---|
Type | Calcium Assay Kit |
Includes: Calcium Indicator, 10X PBX Signal Enhancer
Catalog Number | Mfr. No. | Includes | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Quantity | Content And Storage | Price | Quantity & Availability | |||
BDB640175
|
BD
640175 |
1 vial calcium indicator,10mL PBX signal enhancer (10X, lyophilized), 90mL calcium assay buffer 1X | 10 Preps | -20°C |
Each for $682.00
|
|
|||
BDB640177
|
BD
640177 |
10 vials calcium indicator,100mL PBX signal enhancer (10X, lyophilized) | 100 Preps | Calcium indicator at -20°C and PBX Signal Enhancer 10X at room temperature |
Each for $3,980.00
|
|
|||
Description
- Involves one dye addition step
- Does not require washing
- Allows easy implementation in highthroughput environment
- Probenecid, a standard inhibitor of nonspecific anion transport, is not required for dye-loading
- Can improve retention of the dye in cells
Specifications
For homogeneous measurement of intracellular calcium changes | |
Calcium Assay Kit |
Research use only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title