Learn More
Leica Biosystems BADGE MONITORING FRMALDHY6/CS
Supplier: Leica Biosystems 3801850
Airchek Badges provide an innovative, simple and effective system to monitor vapour levels. Badges are available for formaldehyde, glutaraldehyde and xylene. The Airchek badges provide continuous sampling for personal monitoring and can give the measurements required under Government regulations. The badges are simply clipped on to the technician's clothing and a record is kept of name, location and time. One badge will provide continuous sampling over 8 hours and a second badge can be used to supply information to analyse a 15-minute period during peak exposure.