Learn More
Thomas Scientific Aluminium bottle, UN, AL 99.5, 1200 mL w/ cap
Supplier:  Thomas Scientific 1158W88
Encompass_Preferred
The aluminium bottle is particularly suited for storing, transport or sending substances and reference samples which have to be stored completely unaltered. Other properties: can be completely emptied without residues, odour-neutral, impervious to light, rustproof.
Catalog No. NC1717461