Learn More
Abbott™ iCup™ A.D. Zero Exposure Urine Drug Screens
A zero exposure urine drug screen that tests for up to 12 drugs at once
$280.00 - $663.00
Catalog Number | Mfr. No. | Detectable Analytes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | Price | Quantity & Availability | |||||
22-911-001
|
Abbott™
IDUA147012 |
COC, THC, OPI, mAMP (OX,SG,PH) |
Pack of 25 for $280.00
|
|
|||||
22-911-002
|
Abbott™
IDUA157013 |
COC, THC, OPI, AMP, mAMP (OX, SG, pH) |
Pack of 25 for $321.00
|
|
|||||
22-911-005
|
Abbott™
IDUA167012 |
COC, THC, OPI, AMP, mAMP, PCP (OX, SG, PH) |
Pack of 25 for $388.00
|
|
|||||
22-911-003
|
Abbott™
IDUA157023 |
COC, THC, OPI, AMP, PCP (OX, SG, PH) |
Pack of 25 for $321.00
|
|
|||||
22-911-004
|
Abbott™
IDUA157034 |
COC, THC, OPI, mAMP, PCP (OX, SG, PH) |
Pack of 25 for $321.00
|
|
|||||
22-911-006
|
Abbott™
IDUA167022 |
COC, THC, OPI, AMP, mAMP, BZO (OX, SG, PH) |
Pack of 25 for $388.00
|
|
|||||
22-911-007
|
Abbott™
IDUD187013 |
COC, THC, OPI, AMP, mAMP, PCP, BZO, BAR (OX,SG,PH,NI,GL,CR) |
Pack of 25 for $487.00
|
|
|||||
22-911-009
|
Abbott™
IDUD197014 |
COC, THC, OPI, AMP, mAMP, PCP, BZO, BAR, MTD (OX,SG,PH,NI,GL,CR) |
Pack of 25 for $515.00
|
|
|||||
22-911-011
|
Abbott™
IDUE1107141 |
COC, THC, OPI, AMP, mAMP, BZO, BAR, OXY, MTD, PPX (OX, CR, PH) |
Pack of 25 for $557.00
|
|
|||||
22-911-008
|
Abbott™
IDUE187071 |
COC, THC, MOP, AMP, mAMP, PCP, BZO, OXY(OX,CR,PH) |
Pack of 25 for $487.00
|
|
|||||
22-911-012
|
Abbott™
IDUE1127022 |
COC, THC, OPI, AMP, mAMP, PCP, BZO, BAR, MTD, TCA, OXY, PPX (OX,CR,PH) |
Pack of 25 for $663.00
|
|
|||||
22-911-010
|
Abbott™
IDUD1107012 |
COC, THC, OPI, AMP, mAMP, PCP, BZO, BAR, MTD, TCA (OX,SG,PH,NI,GL,CR) |
Pack of 25 for $557.00
|
|
|||||
Description
Detect the presence of drugs and adulterants in just minutes
- The iCup AD Drug Screen offers versatility with up to 12 drug assays in one cup
- Multiple configurations available
- Built-in validity screening to alleviate tampering concerns
- Room temperature storage
- Comes with temperature strip to verify range of collected sample
- Results can be photocopied for record keeping purposes
- 24-month shelf life
Specifications
Urine | |
iCup™ A.D. Urine Drug Screen | |
25 Pack |