Learn More
3M™ PELTOR™ Lite-Com Pro II UHF Two-Way Radio Communications Headset
Click to view available options
PELTOR Headset
Description
- Ghost voice menu prompts for easy mode selections
- Up to 30 programmable channels in the 403-470 MHz band
- Push-to-Talk (PTT) or hands-free Voice-Activated (VOX) transmit functions
- Rechargeable lithium-ion battery included
- FCC Narrow band mandate compliant
- Environmental microphones on each ear cup offer level-dependent function for situational awareness
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
ANSI S3.1974 | Yes |
For Use With (Equipment) | PELTOR™ Lite-Com Pro II UHF Two-Way Radio Communications Headset |
Headband Cushion | Yes |
Model | MT7H7B4010-NA-50 |
Noise Reduction Rating (NRR) | 26 dB |
Headband Color | Black |
Style | Neckband |