Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Suture Practice Arm and Leg
Simulated limbs with soft vinyl skin over stitchable foam provide a life-like suturing experience for students.
Supplier: 3B Scientific™ W44003
Description
- Realistic skin texture with wrinkles and pores
- Soft and pliable for easy sewing; sutures will not pull out when tightened
- Each limb has three “wounds” that can be sutured repeatedly until surrounding skin wears out; then, new wounds can be cut
- Over 100 cuts can be made on trainer and each can be sutured several times
Specifications
Suture Practice Arm | |
Starter suturing kit | |
2.2 lb. |
Suturing Experience for Students | |
Vinyl, Foam | |
1 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction