Learn More
Cole-Parmer Partlow 711200000021 10" Chart Controller/Recorder; 2 Channel/2 Relay
Supplier: Cole-Parmer 4900192
Proven recording reliability that became the industry standard! Capable of measuring, displaying, recording and controlling up to two process variables from a variety of inputs. All user entered data can be protected against unauthorized changes. Straightforward programming allows you to configure your recorder with a logical step-by-step process using a simple keypad.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.