Learn More
Electron Microscopy Sciences 2 Counting Units, plus Locking Totalizer
SDP
Supplier:  Electron Microscopy Sciences 7260203
Encompass_Preferred
For a quick, accurate count, the durable, long-lasting, ergonomic and easy to use Denominator is the perfect solution for fast, accurate tallies. The counters may be banked or tiered, and various mounting options are available to suit your needs. The Locking Totalizer locks all counters at every 100 counts. A release button quickly unlocks the Denominator for the next 100 counts. Individual percentages can be readily determined by dividing the counter readings by the number of hundreds counted.
Catalog No. 50-192-8548
Explore available promotions