Learn More
Fisher Container™ 1073B Tyvek Autoclavable Precision Clean Breather Bags
$818.00 - $13229.00
Specifications
Certifications/Compliance | NASA |
---|---|
Material | Tyvek, PCHD |
Moisture Vapor Transmission Rate | 1615g/m2/24 hr. and 0.27g/100 in.2/24 hr. |
Thickness (English) | 7.3/3 mil |
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Width (English) | Width (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Width (English) | Width (Metric) | Price | Quantity & Availability | ||
19-351-199
|
Fisher Container™
FISH0038001 |
60 in. | 152.4 cm | 36 in. | 91.4 cm |
Case of 100 for $13,229.00
|
|
||
19-351-190
|
Fisher Container™
91136P |
16 in. | 40.6 cm | 16 in. | 40.6 cm |
Case of 500 for $3,703.00
|
|
||
19-351-191
|
Fisher Container™
FISH0071001 |
21 in. | 53.3 cm | 18 in. | 45.7 cm |
Case of 300 for $2,303.00
|
|
||
19-351-186
|
Fisher Container™
FISH0033001 |
15 in. | 38.1 cm | 12 in. | 30.4 cm |
Case of 1000 for $3,895.00
|
|
||
19-351-192
|
Fisher Container™
GENR0748001 |
24 in. | 60.9 cm | 18 in. | 45.7 cm |
Case of 100 for $818.00
|
|
||
19-351-195
|
Fisher Container™
FISH0070001 |
30 in. | 76.2 cm | 24 in. | 60.9 cm |
Case of 100 for $1,446.00
|
|
||
19-351-184
|
Fisher Container™
91130P |
9 in. | 22.86 cm | 9 in. | 22.8 cm |
Case of 2000 for $7,033.00
|
|
||
19-351-185
|
Fisher Container™
91131P |
12 in. | 30.4 cm | 12 in. | 30.4 cm |
Case of 1000 for $3,026.00
|
|
||
19-351-188
|
Fisher Container™
91134P |
15 in. | 38.1 cm | 15 in. | 38.1 cm |
Case of 500 for $2,653.00
|
|
||
19-351-187
|
Fisher Container™
FISH0032001 |
18 in. | 45.7 cm | 12 in. | 30.4 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
19-351-198
|
Fisher Container™
91144 |
48 in. | 121.9 cm | 36 in. | 91.4 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Specifications
NASA | |
Tyvek, PCHD | |
1615g/m2/24 hr. and 0.27g/100 in.2/24 hr. | |
7.3/3 mil |