Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Pyreneboronic Acid (contains varying amounts of Anhydride), TCI America™
$154.70 - $499.00
Specifications
CAS | 164461-18-1 |
---|---|
Melting Point | 251°C |
Molecular Formula | C16H11BO2 |
Synonym | 1-pyrenylboronic acid, 1-pyreneboronic acid, pyrene-1-boronic acid, 1-boronopyrene, 1-pyrenyl boronic acid |
InChI Key | MWEKPLLMFXIZOC-UHFFFAOYSA-N |
Specifications
164461-18-1 | |
C16H11BO2 | |
MWEKPLLMFXIZOC-UHFFFAOYSA-N | |
pyren-1-ylboronic acid | |
5084102 | |
Crystalline Powder |
251°C | |
1-pyrenylboronic acid, 1-pyreneboronic acid, pyrene-1-boronic acid, 1-boronopyrene, 1-pyrenyl boronic acid | |
B(C1=C2C=CC3=CC=CC4=C3C2=C(C=C1)C=C4)(O)O | |
246.072 | |
246.07 |
Safety and Handling
TSCA : No
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title