Learn More
Usa Safety Solutions LLC 1 CYL BENCH MOUNT BRACKET
Supplier: Usa Safety Solutions LLC GB150FS
Secure one gas cylinder 4 in. to 14 in. in diameter. Posts with swivel feet and non-slip pads clamp to bench or table tops from 1 in. to 2.75 in. thick. Requires 1.5 in. of bench top to secure the bracket. 11 gauge steel. Includes strap. Made in USA.