Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. 1 - 77060360 - 77060360 [-] on 77060360 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-7156-3p | MIMAT0028223 |
Human | hsa-miR-7156-5p | MIMAT0028222 |
Stem-loop Details
Human