Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. 20 - 60564414 - 60564414 [-] on 60564414 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548ag | MIMAT0018969 |
Stem-loop Details
Human