Assay Details
Amplicon Length: | 93 |
Chromosome Location: | Chr. 11 - 47400825 - 47400825 [-] on 47400825 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4487 | MIMAT0019021 |
Stem-loop Details
Human