Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. X - 112780572 - 112780572 [-] on 112780572 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4329 | MIMAT0016923 |
Stem-loop Details
Human