Assay Details
Amplicon Length: | 63 |
Chromosome Location: | Chr. 22 - 35335891 - 35335891 [+] on 35335891 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3909 | MIMAT0018183 |
Stem-loop Details
Human