Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 22 - 21033977 - 21033977 [+] on 21033977 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-649 | MIMAT0003319 |
Stem-loop Details
Human