Assay Details
Target Gene Details
Entrez Gene ID: | 15493 |
Gene Name: | hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2 |
Gene Aliases: |
- |
Location: |
Chr.3:98711107-98724594 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsd3b2 | NM_153193.3 | 6 | 1467 | NP_694873.2 |
BC026757.1 | 6 | 1467 | AAH26757.1 | |
M75886.1 | 1 | 871 |
More Information
Set Membership: |
Intragenic Exonic |