Assay Details
Target Gene Details
Entrez Gene ID: | 22724 |
Gene Name: | zinc finger and BTB domain containing 7B |
Gene Aliases: |
Thpok, Zfp67, c-Krox |
Location: |
Chr.3:89377644-89394814 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zbtb7b | NM_009565.4 | 2 | 1240 | NP_033591.2 |
AK149976.1 | 2 | 1188 | BAE29206.1 | |
AK155784.1 | 2 | 1200 | ||
AK171443.1 | 3 | 1884 | ||
BC046533.1 | 1 | 1079 | AAH46533.1 | |
BC089185.1 | 1 | 874 | AAH89185.1 | |
BC091651.1 | 2 | 1240 | AAH91651.1 |