Assay Details
Target Gene Details
Entrez Gene ID: | 71147 |
Gene Name: | 3-oxoacyl-ACP synthase, mitochondrial |
Gene Aliases: |
4933425A18Rik, C80494 |
Location: |
Chr.14:16238659-16249808 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oxsm | NM_027695.3 | 3 | 2149 | NP_081971.1 |
AK034243.1 | 3 | 1857 | ||
AK083617.1 | 3 | 2150 | BAC38969.1 | |
AK084829.1 | 3 | 1860 | BAC39286.1 |