Assay Details
Target Gene Details
Entrez Gene ID: | 67712 |
Gene Name: | solute carrier family 25, member 37 |
Gene Aliases: |
1700020E22Rik, 4930513O14Rik, 4930526G11Rik, AI848481, C330015G08Rik, Mfrn, Mfrn1, Mscp, frascati, mitoferrin |
Location: |
Chr.14:69241848-69285103 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a37 | NM_026331.3 | 4 | 1237 | NP_080607.2 |
AF288621.1 | 4 | 1190 | AAL23859.1 | |
AF361699.1 | 3 | 2947 | AAL27990.1 | |
AK015770.1 | 4 | 937 | BAB29969.1 | |
AK028588.1 | 2 | 2304 | BAC26021.1 | |
AK046008.1 | 4 | 1613 | ||
AK050414.1 | 4 | 1205 | BAC34243.1 | |
AK162615.1 | 2 | 1099 | BAE36991.1 | |
AK194546.1 | 1 | 137 | ||
BC040399.1 | 2 | 1060 | AAH40399.1 |