Assay Details
Target Gene Details
Entrez Gene ID: | 338371 |
Gene Name: | endonuclease V |
Gene Aliases: |
A730011L01Rik |
Location: |
Chr.11:119491323-119511465 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Endov | NM_001164636.1 | 9 | 2258 | NP_001158108.1 |
NM_177394.3 | 8 | 2310 | NP_796368.1 | |
AK084548.1 | 9 | 2134 | BAC39216.1 | |
AK170595.1 | 8 | 2303 | BAE41901.1 | |
AK172181.1 | 8 | 2277 | BAE42869.1 | |
BC056486.1 | 8 | 2230 | AAH56486.1 |