Assay Details
Target Gene Details
Entrez Gene ID: | 382083 |
Gene Name: | sorting nexin 22 |
Gene Aliases: |
Gm1120 |
Location: |
Chr.9:66065176-66069780 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snx22 | NM_001025612.2 | 7 | 859 | NP_001020783.1 |
AK032028.1 | 7 | 848 | BAC27659.1 | |
BC138950.1 | 7 | 846 | ||
BC138951.1 | 7 | 846 |
More Information
Set Membership: |
Intragenic Exonic |