Assay Details
Target Gene Details
Entrez Gene ID: | 53605 |
Gene Name: | nucleosome assembly protein 1-like 1 |
Gene Aliases: |
AA407126, AI256722, D10Ertd68e, NAP-1 |
Location: |
Chr.10:111473192-111498150 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nap1l1 | NM_001146707.1 | 14 | 3220 | NP_001140179.1 |
AK011301.1 | 1 | 1252 | ||
AK050375.1 | 14 | 3222 | BAC34219.1 | |
BC076591.1 | 14 | 2768 | AAH76591.1 |
More Information
Set Membership: |
Intragenic Exonic |