Assay Details
Target Gene Details
Entrez Gene ID: | 216136 |
Gene Name: | ilvB (bacterial acetolactate synthase)-like |
Gene Aliases: |
5830463I21, AI415009 |
Location: |
Chr.10:78574345-78584502 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ilvbl | NM_173751.4 | 16 | 2039 | NP_776112.1 |
AK030933.1 | 9 | 1664 | ||
AK087956.1 | 15 | 1913 | BAC40057.1 | |
AK146075.1 | 15 | 1967 | BAE26879.1 | |
AK169899.1 | 14 | 2051 | ||
AK184331.1 | 2 | 352 | ||
AK188475.1 | 3 | 266 | ||
AK188806.1 | 3 | 266 | ||
AK197685.1 | 1 | 97 | ||
AK204026.1 | 3 | 266 | ||
AK207520.1 | 3 | 266 | ||
AK207645.1 | 3 | 266 | ||
AK208579.1 | 1 | 85 | ||
AK213840.1 | 3 | 266 | ||
BC056459.1 | 16 | 1997 | AAH56459.1 | |
CK789563.1 | 1 | 310 |
More Information
Set Membership: |
Intragenic Exonic |